Silicon plate, 12.5mm (0.5in) thick, 99.999% (metals basis), Thermo Scientific Chemicals
Silicon plate, 12.5mm (0.5in) thick, 99.999% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Silicon plate, 12.5mm (0.5in) thick, 99.999% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 x 100 mm
25 x 25 mm
50 x 50 mm
Catalog number 039109.GH
also known as 039109-GH
Price (USD)/ Each
1,225.00
-
Add to cart
Quantity:
100 x 100 mm
Request bulk or custom format
Price (USD)/ Each
1,225.00
Add to cart
Silicon plate, 12.5mm (0.5in) thick, 99.999% (metals basis), Thermo Scientific Chemicals
Catalog number039109.GH
Price (USD)/ Each
1,225.00
-
Add to cart
Chemical Identifiers
CAS7440-21-3
IUPAC Namesilicon
Molecular FormulaSi
InChI KeyXUIMIQQOPSSXEZ-UHFFFAOYSA-N
SMILES[Si]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.001% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only