Thermo Scientific Chemicals

Dichloromethane, ACS, 99.5+%, stab. with amylene, Thermo Scientific Chemicals

Catalog number: 039116.AP
500 mL, Each
Thermo Scientific Chemicals

Dichloromethane, ACS, 99.5+%, stab. with amylene, Thermo Scientific Chemicals

Catalog number: 039116.AP
500 mL, Each
Quantity
Catalog number: 039116.AP
also known as 039116-AP
Price (USD)
Have Questions?

Chemical Identifiers

CAS
75-09-2
IUPAC Name
dichloromethane
Molecular Formula
CH2Cl2
InChI Key
YMWUJEATGCHHMB-UHFFFAOYSA-N
SMILES
ClCCl
Assay (GC)
>99.5%
Residual water
≤0.02%
Impurity content
Free halogens P.T.
Free acid (titration)
≤0.0003meq/g
Residue on Evaporation
≤0.002%

Description

Dichloromethane is a chlorinated hydrocarbon that has been employed as a pharamacuetical applications. Mainly, it's utilization as a solvent in manufacturing and food engineering. It is too useful in the drinking bird heat engine. It was the solvent of choice for caffeine extraction. It is likewise employed as a solvent and paint remover. It is useful for the fraction of flash pyrolysis oil and for the extraction of benzo-15-crown-5.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text