Dichloromethane, ACS, 99.5+%, stab. with amylene, Thermo Scientific Chemicals
Dichloromethane, ACS, 99.5+%, stab. with amylene, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Dichloromethane, ACS, 99.5+%, stab. with amylene, Thermo Scientific Chemicals

Maximum level of impurities: Color (APHA) 10, Evaporation residue 0.002%, Titratable acid 0.0003meq/g, Free halogens P.T., H2O 0.02%
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
500 mL
2.5 L
4 x 1 L
4 L
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 039116.K2
also known as 039116-K2
Price (USD)/ Each
48.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
56.90 
Save 8.25 (15%)
-
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (USD)/ Each
48.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
56.90 
Save 8.25 (15%)
Add to cart
Dichloromethane, ACS, 99.5+%, stab. with amylene, Thermo Scientific Chemicals
Catalog number039116.K2
Price (USD)/ Each
48.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
56.90 
Save 8.25 (15%)
-
Add to cart
Chemical Identifiers
CAS75-09-2
IUPAC Namedichloromethane
Molecular FormulaCH2Cl2
InChI KeyYMWUJEATGCHHMB-UHFFFAOYSA-N
SMILESClCCl
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)>99.5%
Residual water≤0.02%
Impurity contentFree halogens P.T.
Free acid (titration)≤0.0003meq/g
Residue on Evaporation≤0.002%
View more
Dichloromethane is a chlorinated hydrocarbon that has been employed as a pharamacuetical applications. Mainly, it's utilization as a solvent in manufacturing and food engineering. It is too useful in the drinking bird heat engine. It was the solvent of choice for caffeine extraction. It is likewise employed as a solvent and paint remover. It is useful for the fraction of flash pyrolysis oil and for the extraction of benzo-15-crown-5.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only