Aluminum lactate, 90-95%, contains ^=5% water, Thermo Scientific Chemicals
Aluminum lactate, 90-95%, contains ^=5% water, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Aluminum lactate, 90-95%, contains ^=5% water, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number 039122.36
also known as 039122-36
Price (USD)/ Each
299.65
Online exclusive
333.00 
Save 33.35 (10%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
299.65
Online exclusive
333.00 
Save 33.35 (10%)
Add to cart
Aluminum lactate, 90-95%, contains ^=5% water, Thermo Scientific Chemicals
Catalog number039122.36
Price (USD)/ Each
299.65
Online exclusive
333.00 
Save 33.35 (10%)
-
Add to cart
Chemical Identifiers
CAS18917-91-4
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormPowder or crystals or crystalline powder
Assay (unspecified)≥90.0%
Water Content≤5.0%
It is used as a skin moisturizer lotion to treat dry, scaly, itchy skin, reagent in biological activity studies of aluminum and in sol-gel processing. Also used in fire foam. Aluminum L-Lactate has been used to inject aluminum into rabbits. Aluminum L-Lactate has been shown to decrease human enamel dissolution in a pH 4 acetate buffer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only