Thermo Scientific Chemicals

Ammonium copper(II) chloride dihydrate, Thermo Scientific Chemicals

Catalog number: 039127.22
100 g, Each
Thermo Scientific Chemicals

Ammonium copper(II) chloride dihydrate, Thermo Scientific Chemicals

Catalog number: 039127.22
100 g, Each
Quantity
Catalog number: 039127.22
also known as 039127-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10060-13-6
IUPAC Name
diammonium tetrachlorocopperdiuide dihydrate
Molecular Formula
Cl4CuH12N2O2
InChI Key
LLMDTWKBBWCWNN-UHFFFAOYSA-L
SMILES
[NH4+].[NH4+].O.O.Cl[Cu--](Cl)(Cl)Cl
Assay (unspecified)
22.9%-23.7% Cu

Description

Ammonium copper(II) chloride dihydrate is used in the determination iron of content in steel. It is also used in coatings, printing ink, rubber, glass, leather and cosmetics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text