Multi-Element QC-19 Check Standard Solution, Specpure™, Thermo Scientific Chemicals
Multi-Element QC-19 Check Standard Solution, Specpure™, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Multi-Element QC-19 Check Standard Solution, Specpure™, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 039146.AE
also known as 039146-AE
Price (USD)/ Each
286.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
337.00 
Save 50.35 (15%)
-
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (USD)/ Each
286.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
337.00 
Save 50.35 (15%)
Add to cart
Multi-Element QC-19 Check Standard Solution, Specpure™, Thermo Scientific Chemicals
Catalog number039146.AE
Price (USD)/ Each
286.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
337.00 
Save 50.35 (15%)
-
Add to cart
SpecificationsSpecification SheetSpecification Sheet
Chemical Name or MaterialMulti-Element QC-19 Check Standard Solution
ConcentrationMatrix: 5% HNO3/tr. F-/tr. tartaric acid
Health Hazard 1H290-H315-H318
Quantity100 mL
Health Hazard 2GHS H Statement
H301-H311-H314-H318-H332
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if inhaled.
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only