Thermo Scientific Chemicals

Boron, plasma standard solution, Specpure™ B 1000μg/mL, Thermo Scientific Chemicals

Catalog number: 039147.AE
100 mL, Each
Thermo Scientific Chemicals

Boron, plasma standard solution, Specpure™ B 1000μg/mL, Thermo Scientific Chemicals

Catalog number: 039147.AE
100 mL, Each
Quantity
Catalog number: 039147.AE
also known as 039147-AE
Price (USD)
Quantity
-
Have Questions?
Concentration
1,000?g/mL ?1%
Starting Material
H₃BO₃
Concentration
1,000 ? 3 ?g/ml Boron
Matrix
H₂O

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text