Thermo Scientific Chemicals

Potassium ethoxide, 99+% (metals basis), 15% w/v in ethanol, Thermo Scientific Chemicals

Catalog number: 039161.AC
25 mL, Each
Thermo Scientific Chemicals

Potassium ethoxide, 99+% (metals basis), 15% w/v in ethanol, Thermo Scientific Chemicals

Catalog number: 039161.AC
25 mL, Each
Quantity
Catalog number: 039161.AC
also known as 039161-AC
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
917-58-8
Appearance (Color)
Pale yellow
Gravimetric analysis
≥ 99%
Trace Metals (by ICP)
< 5000ppm
Form
Liquid

Description

Potassium ethoxide is used widely among others to alkaline varnish degradation . Potassium ethoxide is used as a strong base. It is used to perform transesterification reactions that yield ethyl esters. It is also a suitable base for the malonic ester synthesis. It is also used as Pharmaceutical Intermediates. Other applications include Nano-materials & Thin Films, Engineering & Environmental, Catalysis & Synthesis, Healthcare & Biochemical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For Manufacturing and Laboratory Use. Not for beverage use

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text