Thermo Scientific Chemicals

Potassium isopropoxide, 99% (metals basis), 5% w/v in isopropanol, Thermo Scientific Chemicals

Catalog number: 039162.AE
100 mL, Each
Thermo Scientific Chemicals

Potassium isopropoxide, 99% (metals basis), 5% w/v in isopropanol, Thermo Scientific Chemicals

Catalog number: 039162.AE
100 mL, Each
Quantity
Catalog number: 039162.AE
also known as 039162-AE
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6831-82-9
IUPAC Name
potassium propan-2-olate
Molecular Formula
C3H7KO
InChI Key
WQKGAJDYBZOFSR-UHFFFAOYSA-N
SMILES
[K+].CC(C)[O-]
Appearance (Color)
Clear colorless
Form
Liquid
Chloride content
≤100ppm
Concentration (w/v)
4.5-5.5%
Total Metal Impurities
≤1%

Description

It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals. They are used in wide range of applications such as colorants and aroma chemicals. They are used in manufacturing detergents and biodiesel. They also act as catalysts in polymerization and isomerizations. Other uses include used for formation of ethers. Also acts as a moderately strong base for deprotonation and base catalysed reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text