Specifications
Chemical Name or MaterialTin(IV) 2-ethylhexanoate diisopropoxide
CAS288374-32-3
Health Hazard 1H315-H319-H361d
Health Hazard 2GHS H Statement
H361-H315-H319
Suspected of damaging fertility or the unborn child.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3P201-P202-P264b-P280i-P281-P302+P352-P305+P351+P338-P308+P313-P332+P313-P362-P501c
View more