Thermo Scientific Chemicals

Tungsten(VI) ethoxide, 99.8% (metals basis), 5% w/v in ethanol, Thermo Scientific Chemicals

Catalog number: 039170.AE
100 mL, Each
Thermo Scientific Chemicals

Tungsten(VI) ethoxide, 99.8% (metals basis), 5% w/v in ethanol, Thermo Scientific Chemicals

Catalog number: 039170.AE
100 mL, Each
Quantity
Catalog number: 039170.AE
also known as 039170-AE
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
62571-53-3
IUPAC Name
hexaethoxytungsten
Molecular Formula
C12H30O6W
InChI Key
JHZFYDXXWXXVRM-UHFFFAOYSA-N
SMILES
CCO[W](OCC)(OCC)(OCC)(OCC)OCC
Impurity content
Al: ≤10ppm
Impurity content
B: ≤10ppm
Impurity content
Ca: ≤5ppm
Impurity content
Fe: ≤30ppm
Impurity content
Nb: ≤10ppm

Description

Tungsten(VI) ethoxide is used in nano-materials, thin films, display and imaging, optics and glasses. It is also used for x-ray screen phosphors, fireproofing fabrics and in gas sensors. It acts as an intermediate for sol-gel coatings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For Manufacturing and Laboratory Use. Not for beverage use

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text