Thermo Scientific Chemicals

Zirconium(IV) 2-ethylhexanoate, 97%, Thermo Scientific Chemicals

Catalog number: 039174.14
25 g, Each
Thermo Scientific Chemicals

Zirconium(IV) 2-ethylhexanoate, 97%, Thermo Scientific Chemicals

Catalog number: 039174.14
25 g, Each
Quantity
Catalog number: 039174.14
Price (USD)
Have Questions?

Chemical Identifiers

CAS
2233-42-3
IUPAC Name
zirconium(4+) tetrakis(2-ethylhexanoate)
Molecular Formula
C32H60O8Zr
InChI Key
OFYFURKXMHQOGG-UHFFFAOYNA-J
SMILES
[Zr+4].CCCCC(CC)C([O-])=O.CCCCC(CC)C([O-])=O.CCCCC(CC)C([O-])=O.CCCCC(CC)C([O-])=O
Form
Viscous liquid
Appearance (Color)
Clear yellow
Assay from Supplier's CofA
≥96.0%

Description

Zirconium(IV) 2-ethylhexanoate is utilized as a metal organic precursor of zirconium for solution deposition and chemical vapor deposition of thin films containing zirconium especially zirconium oxide thin films. It is also used as yttria stabilized zirconia films for solid oxide fuel cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text