Thermo Scientific Chemicals

1-Methyl-2-pyrrolidinone, Spectrophotometric Grade, 99.5+%, Thermo Scientific Chemicals

Catalog number: 039176.M1
4 x 1 L, Each
Thermo Scientific Chemicals

1-Methyl-2-pyrrolidinone, Spectrophotometric Grade, 99.5+%, Thermo Scientific Chemicals

Catalog number: 039176.M1
4 x 1 L, Each
Quantity
Catalog number: 039176.M1
also known as 039176-M1
Price (USD)
Have Questions?

Chemical Identifiers

CAS
872-50-4
IUPAC Name
1-methylpyrrolidin-2-one
Molecular Formula
C5H9NO
InChI Key
SECXISVLQFMRJM-UHFFFAOYSA-N
SMILES
CN1CCCC1=O
Residual water
0.05%
Residue on Evaporation
6ppm

Description

1-methyl-2-pyrrolidinone, is useful for spectrophotometry, chromatography and ICP-MS detection. It is useful for the chemical functionalization of graphene sheets by solvothermal reduction of graphene oxide. It is also useful for pharmaceutical and different industrial applications due to its broad solvency and low volatility.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Maximum level of impurities: Evaporation residue 6ppm, H2O 0.05%
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text