Thulium(III) bromide, ultra dry, 99.99% (REO), Thermo Scientific Chemicals
Thulium(III) bromide, ultra dry, 99.99% (REO), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Thulium(III) bromide, ultra dry, 99.99% (REO), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number 039186.06
also known as 039186-06
Price (USD)/ Each
574.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
574.00
Add to cart
Thulium(III) bromide, ultra dry, 99.99% (REO), Thermo Scientific Chemicals
Catalog number039186.06
Price (USD)/ Each
574.00
-
Add to cart
Chemical Identifiers
CAS14456-51-0
IUPAC Namethulium(3+) tribromide
Molecular FormulaBr3Tm
InChI KeyHQSWGSFQSCMHFQ-UHFFFAOYSA-K
SMILES[Br-].[Br-].[Br-].[Tm+3]
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Total Rare Earth Oxide Impurities0.01% max.
Appearance (Color)Yellow
Residual water< 100 ppm
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Ampouled under argon
RUO – Research Use Only