n-Butyl acetate, HPLC Grade, 99.5+%, Thermo Scientific Chemicals
n-Butyl acetate, HPLC Grade, 99.5+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

n-Butyl acetate, HPLC Grade, 99.5+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
4 L
250 mL
1 L
4 x 1 L
Catalog number 039197.K7
also known as 039197-K7
Price (USD)/ Each
260.00
-
Add to cart
Quantity:
4 L
Request bulk or custom format
Price (USD)/ Each
260.00
Add to cart
n-Butyl acetate, HPLC Grade, 99.5+%, Thermo Scientific Chemicals
Catalog number039197.K7
Price (USD)/ Each
260.00
-
Add to cart
Chemical Identifiers
CAS123-86-4
IUPAC Namebutyl acetate
Molecular FormulaC6H12O2
InChI KeyDKPFZGUDAPQIHT-UHFFFAOYSA-N
SMILESCCCCOC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Optical Absorbance at254 nm: ≤1.00 AU
Optical Absorbance at259 nm: ≤0.20
Optical Absorbance at300 nm: ≤0.02 AU
Optical Absorbance at275 nm: ≤0.04 AU
Color (APHA)≤10
View more
n-Butyl acetate is used as a mobile phase in High Performance Liquid Chromatography and Liquid Chromatography coupled with Mass Spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only