1-Chlorobutane, HPLC Grade, 99.5+%, Thermo Scientific Chemicals
1-Chlorobutane, HPLC Grade, 99.5+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Chlorobutane, HPLC Grade, 99.5+%, Thermo Scientific Chemicals

Maximum level of impurities: Evaporation residue 1ppm, H2O 0.02%
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
250 mL
4 x 1 L
4 L
Catalog number 039198.K2
also known as 039198-K2
Price (USD)/ Each
91.65
Online exclusive
102.00 
Save 10.35 (10%)
-
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (USD)/ Each
91.65
Online exclusive
102.00 
Save 10.35 (10%)
Add to cart
1-Chlorobutane, HPLC Grade, 99.5+%, Thermo Scientific Chemicals
Catalog number039198.K2
Price (USD)/ Each
91.65
Online exclusive
102.00 
Save 10.35 (10%)
-
Add to cart
Chemical Identifiers
CAS109-69-3
IUPAC Name1-chlorobutane
Molecular FormulaC4H9Cl
InChI KeyVFWCMGCRMGJXDK-UHFFFAOYSA-N
SMILESCCCCCl
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥99.5%
Optical Absorbance at220nm: ≤1.00 AU
Optical Absorbance at225nm: ≤0.50 AU
Optical Absorbance at400nm: ≤0.005 AU
Water Content (Karl Fischer Titration)≤0.02%
View more
1-Chlorobutane is used as intermediates and solvents. It is used for HPLC, spectrophotometry and environmental testing. It is used as alkylating reagent in organic and organometallic compound synthesis. It is also employed as an antihelmintic in veterinary medicine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only