n-Hexane, HPLC Grade, 95% min, Thermo Scientific Chemicals
n-Hexane, HPLC Grade, 95% min, Thermo Scientific Chemicals
Thermo Scientific Chemicals

n-Hexane, HPLC Grade, 95% min, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
4 x 1 L
1 L
4 L
Catalog number 039199.M1
also known as 039199-M1
Price (USD)/ Each
198.65
Online exclusive
221.00 
Save 22.35 (10%)
-
Add to cart
Quantity:
4 x 1 L
Request bulk or custom format
Price (USD)/ Each
198.65
Online exclusive
221.00 
Save 22.35 (10%)
Add to cart
n-Hexane, HPLC Grade, 95% min, Thermo Scientific Chemicals
Catalog number039199.M1
Price (USD)/ Each
198.65
Online exclusive
221.00 
Save 22.35 (10%)
-
Add to cart
Chemical Identifiers
CAS110-54-3
IUPAC Namehexane
Molecular FormulaC6H14
InChI KeyVLKZOEOYAKHREP-UHFFFAOYSA-N
SMILESCCCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥95.0%
CommentSubmicron filtered
FormLiquid
Appearance (Color)Colorless
Water Content (Karl Fischer Titration)0.03%, max
View more
as a solvent to extract edible oils from seed and vegetable crops. Commercial grades of hexane are used as solvents for glues, varnishes, and inks, cleaning agent (degreaser) in the printing industry, liquid in low temperature thermometers. other uses are as Fuels and fuel additives, Intermediates and Laboratory chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only