Thermo Scientific Chemicals

Caffeine, 99.7%, Thermo Scientific Chemicals

Catalog number: 039214.22
100 g, Each
Thermo Scientific Chemicals

Caffeine, 99.7%, Thermo Scientific Chemicals

Catalog number: 039214.22
100 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: 039214.22
also known as 039214-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
58-08-2
IUPAC Name
1,3,7-trimethyl-2,3,6,7-tetrahydro-1H-purine-2,6-dione
Molecular Formula
C8H10N4O2
InChI Key
RYYVLZVUVIJVGH-UHFFFAOYSA-N
SMILES
CN1C=NC2=C1C(=O)N(C)C(=O)N2C
Residue on ignition
0.1%, max
Loss on Drying
0.5%, max
Appearance (Color)
White to off-white
Form
Powder
Assay (Titration)
>/= 99.6% ( on dry basis)

Description

It acts as a stimulant drug and a reversible acetylcholinesterase inhibitor. It plays an important role for the inactivation of Escherichia coli O 157:H 7 in brain heart infusion (BHI) broth. It is also useful for the protection from photodamage after ultraviolet irradiation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text