Cerium(III) phosphate, 99% min, Thermo Scientific Chemicals
Cerium(III) phosphate, 99% min, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cerium(III) phosphate, 99% min, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Catalog number 039215.36
also known as 039215-36
Price (USD)/ Each
428.65
Online exclusive
476.00 
Save 47.35 (10%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
428.65
Online exclusive
476.00 
Save 47.35 (10%)
Add to cart
Cerium(III) phosphate, 99% min, Thermo Scientific Chemicals
Catalog number039215.36
Price (USD)/ Each
428.65
Online exclusive
476.00 
Save 47.35 (10%)
-
Add to cart
Chemical Identifiers
CAS13454-71-2
IUPAC Namecerium(3+) phosphate
Molecular FormulaCeO4P
InChI KeyTYAVIWGEVOBWDZ-UHFFFAOYSA-K
SMILES[Ce+3].[O-]P([O-])([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)98.6% min
Intermediate product in cerium-based technique for staining lysosomal enzymes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only