Calcium arsenate, 95%, Thermo Scientific Chemicals
Calcium arsenate, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Calcium arsenate, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Packaging:
Packing group II
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 039219.18
also known as 039219-18
Price (USD)/ Each
87.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
103.00 
Save 15.35 (15%)
-
Add to cart
Quantity:
50 g
Packaging:
Packing group II
Request bulk or custom format
Price (USD)/ Each
87.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
103.00 
Save 15.35 (15%)
Add to cart
Calcium arsenate, 95%, Thermo Scientific Chemicals
Catalog number039219.18
Price (USD)/ Each
87.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
103.00 
Save 15.35 (15%)
-
Add to cart
Chemical Identifiers
CAS7778-44-1
IUPAC Nametricalcium diarsorate
Molecular FormulaAs2Ca3O8
InChI KeyRMBBSOLAGVEUSI-UHFFFAOYSA-H
SMILES[Ca++].[Ca++].[Ca++].[O-][As]([O-])([O-])=O.[O-][As]([O-])([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)94.1% min
Calcium arsenate is also used in weather- resistant wood treatment. It was used mainly in viticulture, in the past. it was mainly used against the boll weevil in cotton cultivation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only