Hafnium telluride, 99.5% (metals basis), Thermo Scientific Chemicals
Hafnium telluride, 99.5% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Hafnium telluride, 99.5% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 039223.03
also known as 039223-03
Price (USD)/ Each
134.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
158.00 
Save 23.35 (15%)
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
134.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
158.00 
Save 23.35 (15%)
Add to cart
Hafnium telluride, 99.5% (metals basis), Thermo Scientific Chemicals
Catalog number039223.03
Price (USD)/ Each
134.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
158.00 
Save 23.35 (15%)
-
Add to cart
Chemical Identifiers
CAS39082-23-0
IUPAC Nameditellanylidenehafnium
Molecular FormulaHfTe2
InChI KeyBCMXGUYIVFZMCT-UHFFFAOYSA-N
SMILES[Te]=[Hf]=[Te]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.5% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only