Thermo Scientific Chemicals

Bis(1,5-cyclooctadiene)nickel(0), Thermo Scientific Chemicals

Catalog number: 039234.09
10 g, Each
Thermo Scientific Chemicals

Bis(1,5-cyclooctadiene)nickel(0), Thermo Scientific Chemicals

Catalog number: 039234.09
10 g, Each
Quantity
Catalog number: 039234.09
also known as 039234-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1295-35-8
IUPAC Name
bis((1Z,5Z)-cycloocta-1,5-diene) nickel
Molecular Formula
C16H24Ni
InChI Key
JRTIUDXYIUKIIE-KZUMESAESA-N
SMILES
[Ni].C1C\C=C/CC\C=C/1.C1C\C=C/CC\C=C/1
Gravimetric analysis
Ni=20.9-21.8 wt%
Appearance (Color)
Yellow to orange.
Form
Crystalline powder.
Gravimetric analysis
>98.0%

Description

Bis(1,5-cyclooctadiene)nickel(0) is used as a catalyst for the cycloaddition of 1,3-dienes and is used to catalyze the addition of allyl phenyl sulfide to alkynes leading to 1,4-dienes. Also, it is involved as a catalyst for asymmetric alpha-arylation and heteroarylation of ketones with chloroarenes, stereoselective borylative ketone-diene coupling, cycloaddition of benzamides with internal alkynes and methyl carboxylation of homopropargylic alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text