Lanthanum aluminum oxide, 99.9% (metals basis), Thermo Scientific Chemicals
Lanthanum aluminum oxide, 99.9% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Lanthanum aluminum oxide, 99.9% (metals basis), Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number 039250.14
also known as 039250-14
Price (USD)/ Each
181.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
181.00
Add to cart
Lanthanum aluminum oxide, 99.9% (metals basis), Thermo Scientific Chemicals
Catalog number039250.14
Price (USD)/ Each
181.00
-
Add to cart
Chemical Identifiers
CAS12003-65-5
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.1% max.
Used in epitaxial thin films, non-epitaxial thin films and substrate. Mainly used in making optical glass parts and optical fibers for electronic industry. Also often used in ceramic industry and catalysts. Due to its dielectric properties it is also posses properties for low-loss microwave and dielectric resonance applications.