Ferrocenecarboxaldehyde, 97%, Thermo Scientific Chemicals
Ferrocenecarboxaldehyde, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ferrocenecarboxaldehyde, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
2 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 039254.09
also known as 039254-09
Price (USD)/ Each
-
Quantity:
10 g
Request bulk or custom format
Chemical Identifiers
CAS12093-10-6
Molecular FormulaC11H10FeO
SMILES[Fe].c1cccc1.O=Cc1cccc1
Molecular Weight (g/mol)214.05
MDL NumberMFCD00001429
SpecificationsSpecification SheetSpecification Sheet
FormPowder or crystals or crystalline powder
Assay from Suppliers CofA≥96.0%
Ferrocenecarboxaldehyde, is used to prepare chiral ferrocene aziridinylmethanols for selective azomethine ylide cycloaddtion. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only