Thermo Scientific Chemicals

Iron(III) hexacyanoferrate(II), Thermo Scientific Chemicals

Catalog number: 039259.14
25 g, Each
Thermo Scientific Chemicals

Iron(III) hexacyanoferrate(II), Thermo Scientific Chemicals

Catalog number: 039259.14
25 g, Each
Quantity
Catalog number: 039259.14
also known as 039259-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
14038-43-8
IUPAC Name
tetrairon(3+) tris(λ²-iron(2+)) octadecakis(iminomethanide)
Molecular Formula
C18Fe7N18
InChI Key
DCYOBGZUOMKFPA-UHFFFAOYSA-N
SMILES
[Fe++].[Fe++].[Fe++].[Fe+3].[Fe+3].[Fe+3].[Fe+3].[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N
Assay (unspecified)
31% Fe min

Description

Iron(III) hexacyanoferrate(II) is uses extensively as an Iron Oxide dye. Prussian blue is used as a paint and wallpaper printing, chemical coatings, carbon paper and in the plastics industry. as an antidote for poisoning with radioactive cesium or thallium. In the metalworking and mechanical engineering Prussian blue is thinly applied as a paste on metal surfaces in order to assess the quality scraped surfaces can.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text