Thermo Scientific Chemicals

Iron(II) oxalate hydrate, 96%, Thermo Scientific Chemicals

Catalog number: 039261.A3
2 kg, Each
Thermo Scientific Chemicals

Iron(II) oxalate hydrate, 96%, Thermo Scientific Chemicals

Catalog number: 039261.A3
2 kg, Each
Quantity
Catalog number: 039261.A3
also known as 039261-A3
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
516-03-0
IUPAC Name
λ²-iron(2+) oxalate
Molecular Formula
C2FeO4
InChI Key
OWZIYWAUNZMLRT-UHFFFAOYSA-L
SMILES
[Fe++].[O-]C(=O)C([O-])=O
Form
Powder
Appearance (Color)
Yellow
Assay from Supplier's CofA
≥95.0 to ≤105.0% (Redox titration)

Description

Used as a photographic developer, glass tint, decorative glass colorant, and pigment in plastics, paints, and lacquers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text