1x1 image pixel for data collection
Benzyldiphenylphosphine, 99%
Benzyldiphenylphosphine, 99%
Benzyldiphenylphosphine, 99%
Benzyldiphenylphosphine, 99%
Thermo Scientific Chemicals

Benzyldiphenylphosphine, 99%

CAS: 7650-91-1 | C19H17P | 276.32 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
0.5 g
2 g
10 g
Catalog number 039267.02
also known as 039267-02
Price (USD)
30.80
Each
Add to cart
Quantity:
0.5 g
Request bulk or custom format
Price (USD)
30.80
Each
Add to cart
Specifications
CAS7650-91-1
Chemical Name or MaterialBenzyldiphenylphosphine
Formula Weight276.32
Health Hazard 1H302-H315-H319-H335
Health Hazard 3P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
View more
Employed as a catalyst for Suzuki cross-coupling and antiarthritic agent (inhibitor of cathepsin B). It is also used as catalyst in pharmaceutical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Share catalog number, name or link