Bis(dimethylphosphino)methane, 98%, Thermo Scientific Chemicals
Bis(dimethylphosphino)methane, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Bis(dimethylphosphino)methane, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
0.5 g
Catalog number 039275.04
also known as 039275-04
Price (USD)/ Each
291.65
Online exclusive
324.00 
Save 32.35 (10%)
-
Add to cart
Quantity:
2 g
Request bulk or custom format
Price (USD)/ Each
291.65
Online exclusive
324.00 
Save 32.35 (10%)
Add to cart
Bis(dimethylphosphino)methane, 98%, Thermo Scientific Chemicals
Catalog number039275.04
Price (USD)/ Each
291.65
Online exclusive
324.00 
Save 32.35 (10%)
-
Add to cart
Specifications
Chemical Name or MaterialBis(dimethylphosphino)methane
CAS64065-08-3
Health Hazard 1H250-H252-H315-H319-H335
Health Hazard 2GHS H Statement
H250-H252-H315-H319-H335
Catches fire spontaneously if exposed to air.
Self-heating in large quantities
may catch fire.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P210-P222-P235+P410-P261-P264b-P271-P280-P302+P334-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Packaged under argon
RUO – Research Use Only