Dichlorobis(ethylenediamine)platinum(II), Pt 50%, Thermo Scientific Chemicals
Dichlorobis(ethylenediamine)platinum(II), Pt 50%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Dichlorobis(ethylenediamine)platinum(II), Pt 50%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
0.25 g
1 g
Catalog number 039280.01
also known as 039280-01
Price (USD)/ Each
78.30
-
Add to cart
Quantity:
0.25 g
Request bulk or custom format
Price (USD)/ Each
78.30
Add to cart
Dichlorobis(ethylenediamine)platinum(II), Pt 50%, Thermo Scientific Chemicals
Catalog number039280.01
Price (USD)/ Each
78.30
-
Add to cart
Specifications
Chemical Name or MaterialDichlorobis(ethylenediamine)platinum(II)
CAS21430-85-3
Health Hazard 1H315-H317-H319-H334-H335
Health Hazard 2GHS H Statement
H334-H335-H315-H319-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Health Hazard 3P261-P264b-P271-P272-P280-P285-P302+P352-P304+P340-P305+P351+P338-P333+P313-P342+P311-P362-P501c
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only