Specifications
Chemical Name or MaterialDi-mu-chlorodichlorobis(ethylene)diplatinum(II)
CAS12073-36-8
Health Hazard 1H315-H317-H319-H334-H335
Health Hazard 2GHS H Statement
H334-H335-H315-H319-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Health Hazard 3P261-P264b-P271-P272-P280-P285-P302+P352-P304+P340-P305+P351+P338-P333+P313-P342+P311-P362-P501c
View more