Thermo Scientific Chemicals

Iodotrimethylplatinum(IV), Thermo Scientific Chemicals

Catalog number: 039286.02
0.5 g, Each
Thermo Scientific Chemicals

Iodotrimethylplatinum(IV), Thermo Scientific Chemicals

Catalog number: 039286.02
0.5 g, Each
Quantity
Catalog number: 039286.02
also known as 039286-02
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
14364-93-3
IUPAC Name
iodoplatinum trimethanide
Molecular Formula
C3H9IPt
InChI Key
ZCSQPOLLUOLHHF-UHFFFAOYSA-M
SMILES
[CH3-].[CH3-].[CH3-].I[Pt]
Assay (unspecified)
99% min

Description

They are used as catalysts in organic reactions. It is used in chemical, organic Intermediates research. They are used as platinum group compounds for the preparation of materials suitable for industrial applications, mainly in electronics as coatings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text