Bis(1,5-cyclooctadiene)rhodium(I) trifluoromethanesulfonate, Thermo Scientific Chemicals
Bis(1,5-cyclooctadiene)rhodium(I) trifluoromethanesulfonate, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Bis(1,5-cyclooctadiene)rhodium(I) trifluoromethanesulfonate, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mg
2 g
Catalog number 039289.ME
also known as 039289-ME
Price (USD)/ Each
-
Quantity:
500 mg
Request bulk or custom format
Chemical Identifiers
CAS99326-34-8
IUPAC Nameλ¹-rhodium(1+) bis((1Z,5Z)-cycloocta-1,5-diene) trifluoromethanesulfonate
Molecular FormulaC17H24F3O3RhS
InChI KeyGOPBTCMAUMSOBX-QMDOQEJBSA-M
SMILES[Rh+].[O-]S(=O)(=O)C(F)(F)F.C1C\C=C/CC\C=C/1.C1C\C=C/CC\C=C/1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)98% min
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Packaged under argon
RUO – Research Use Only