Thermo Scientific Chemicals

Chlorobis(ethylene)rhodium(I) dimer, 99%, Thermo Scientific Chemicals

Catalog number: 039291.03
1 g, Each
Thermo Scientific Chemicals

Chlorobis(ethylene)rhodium(I) dimer, 99%, Thermo Scientific Chemicals

Catalog number: 039291.03
1 g, Each
Quantity
Catalog number: 039291.03
also known as 039291-03
Price (USD)
Have Questions?

Chemical Identifiers

CAS
12081-16-2
IUPAC Name
bis(chlororhodium); tetrakis(ethene)
Molecular Formula
C8H16Cl2Rh2
InChI Key
OOQJCPOXJFCGCR-UHFFFAOYSA-L
SMILES
Cl[Rh].Cl[Rh].C=C.C=C.C=C.C=C
Assay (unspecified)
98.6% min

Description

Chlorobis(ethylene)rhodium(I) dimer is used as a precursor to rhodium(I) norbornadiene catalyst, which is used in an asymmetric 1,4-addition of arylboronic acids to cyclic enones. Further, it serves as an important organic intermediate. It catalyzes the dimerization of ethylene to 1-butene. It is also used in the preparation of homogeneous catalysts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text