Thermo Scientific Chemicals

Dicarbonyl(2,4-pentanedionato)rhodium(I), 97%, Thermo Scientific Chemicals

Catalog number: 039295.06
5 g, Each
Thermo Scientific Chemicals

Dicarbonyl(2,4-pentanedionato)rhodium(I), 97%, Thermo Scientific Chemicals

Catalog number: 039295.06
5 g, Each
Quantity
Catalog number: 039295.06
also known as 039295-06
Price (USD)
Have Questions?

Chemical Identifiers

CAS
14874-82-9
IUPAC Name
λ¹-rhodium(1+) bis(methanidylidyneoxidanium) (2Z)-4-oxopent-2-en-2-olate
Molecular Formula
C7H7O4Rh
InChI Key
BZCAWKOPWNIDOC-FGSKAQBVSA-M
SMILES
[Rh+].[C-]#[O+].[C-]#[O+].C\C([O-])=C\C(C)=O
Assay (unspecified)
≤96.0%
Appearance (Color)
Green
Form
Crystalline powder or powder

Description

(Acetylacetonato)dicarbonylrhodium(I) is used in in situ formation of a floors-soluble hydroformylation catalyst of interest in molecular engineering. It is also used as a catalyst for various carbonylation reactions, silylcarbocyclizations, conjugate additions to eons, carbamoylstannation, and reduction of aromatic nitro compounds. It is also used in Hydroformylation reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text