Pentamethylcyclopentadienylmolybdenum dicarbonyl dimer, 99%, Thermo Scientific Chemicals
Pentamethylcyclopentadienylmolybdenum dicarbonyl dimer, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Pentamethylcyclopentadienylmolybdenum dicarbonyl dimer, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number 039303.MD
also known as 039303-MD
Price (USD)/ Each
46.20
-
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (USD)/ Each
46.20
Add to cart
Pentamethylcyclopentadienylmolybdenum dicarbonyl dimer, 99%, Thermo Scientific Chemicals
Catalog number039303.MD
Price (USD)/ Each
46.20
-
Add to cart
Specifications
Chemical Name or MaterialPentamethylcyclopentadienylmolybdenum dicarbonyl dimer
CAS12132-04-6
Health Hazard 1H300+H330
Health Hazard 2GHS H Statement
H300-H330
Fatal if swallowed.
Fatal if inhaled.
Health Hazard 3P260-P264b-P270-P271-P284-P301+P310-P304+P340-P310-P330-P501c
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only