Thermo Scientific Chemicals
Mercury(II) trifluoromethanesulfonate, 98%, Thermo Scientific Chemicals
Catalog number: 039305.14
25 g, Each
Thermo Scientific Chemicals
Mercury(II) trifluoromethanesulfonate, 98%, Thermo Scientific Chemicals
Catalog number: 039305.14
25 g, Each
Quantity
Catalog number: 039305.14
also known as 039305-14
Price (USD)
Have Questions?
Description
Hg(OTf)2 exhibits remarkable catalytic activity for the hydroxylative cyclization of 1,6-enynes. Mercuric triflate is a very versatile reagent and has been used for several organic catalytic transformations including C-C bond forming cyclizations, alkyne hydrations, heterocycle synthesis and very recently in C-N bond forming reactions. Commonly used as the metal source for Lewis acid catalyzed asymmetric reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov