Specifications
Chemical Name or MaterialAluminum magnesium isopropoxide
CAS69207-83-6
Health Hazard 1H228-H315-H319-H335
Health Hazard 2GHS H Statement
H225-H228-H315-H319-H335
Highly flammable liquid and vapor.
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
View more