Aluminum magnesium isopropoxide, 99.95+% (metals basis), Thermo Scientific Chemicals
Aluminum magnesium isopropoxide, 99.95+% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Aluminum magnesium isopropoxide, 99.95+% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
2 g
50 g
Catalog number 039320.09
also known as 039320-09
Price (USD)/ Each
191.00
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
191.00
Add to cart
Aluminum magnesium isopropoxide, 99.95+% (metals basis), Thermo Scientific Chemicals
Catalog number039320.09
Price (USD)/ Each
191.00
-
Add to cart
Specifications
Chemical Name or MaterialAluminum magnesium isopropoxide
CAS69207-83-6
Health Hazard 1H228-H315-H319-H335
Health Hazard 2GHS H Statement
H225-H228-H315-H319-H335
Highly flammable liquid and vapor.
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only