Lithium cyclopentadienide, 97%, Thermo Scientific Chemicals
Lithium cyclopentadienide, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Lithium cyclopentadienide, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 039329.06
also known as 039329-06
Price (USD)/ Each
64.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
64.00
Add to cart
Lithium cyclopentadienide, 97%, Thermo Scientific Chemicals
Catalog number039329.06
Price (USD)/ Each
64.00
-
Add to cart
Chemical Identifiers
CAS16733-97-4
IUPAC Namelithium(1+) cyclopenta-1,3-dien-1-ide
Molecular FormulaC5H5Li
InChI KeyCSOHRTAOCDVTQU-UHFFFAOYSA-N
SMILES[Li+].C1C=CC=[C-]1
View more
SpecificationsSpecification SheetSpecification Sheet
Formpowder
Appearance (Color)White to off-white
Assay (Titration)> 96.0%
Lithium cyclopentadienide is a useful reagent in preparing metallocenes and cyclopentadienyl derivatives. It is also used as intermediates and in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only