Lithium salicylate, tech., Thermo Scientific Chemicals
Lithium salicylate, tech., Thermo Scientific Chemicals
Thermo Scientific Chemicals

Lithium salicylate, tech., Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number 039356.14
also known as 039356-14
Price (USD)/ Each
37.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
37.00
Add to cart
Lithium salicylate, tech., Thermo Scientific Chemicals
Catalog number039356.14
Price (USD)/ Each
37.00
-
Add to cart
Specifications
Chemical Name or MaterialLithium salicylate
CAS552-38-5
Health Hazard 1H302-H314-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only