Magnesium niobium oxide, 99.9% (metals basis), Thermo Scientific Chemicals
Magnesium niobium oxide, 99.9% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Magnesium niobium oxide, 99.9% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
250 g
Catalog number 039373.18
also known as 039373-18
Price (USD)/ Each
372.00
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
372.00
Add to cart
Magnesium niobium oxide, 99.9% (metals basis), Thermo Scientific Chemicals
Catalog number039373.18
Price (USD)/ Each
372.00
-
Add to cart
Chemical Identifiers
CAS12163-26-7
IUPAC Namediniobium(5+) dimagnesium(2+) heptaoxidandiide
Molecular FormulaMg2Nb2O7
InChI KeyCWMQWEHVVUJOBW-UHFFFAOYSA-N
SMILES[O--].[O--].[O--].[O--].[O--].[O--].[O--].[Mg++].[Mg++].[Nb+5].[Nb+5]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.1% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only