Manganese(II) tungsten oxide, 99.9% (metals basis), Thermo Scientific Chemicals
Manganese(II) tungsten oxide, 99.9% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Manganese(II) tungsten oxide, 99.9% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number 039374.18
also known as 039374-18
Price (USD)/ Each
148.00
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
148.00
Add to cart
Manganese(II) tungsten oxide, 99.9% (metals basis), Thermo Scientific Chemicals
Catalog number039374.18
Price (USD)/ Each
148.00
-
Add to cart
Chemical Identifiers
CAS14177-46-9
IUPAC Namemanganese(2+) dioxotungstenbis(olate)
Molecular FormulaMnO4W
InChI KeyCRLHSBRULQUYOK-UHFFFAOYSA-N
SMILES[Mn++].[O-][W]([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.1% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only