Thermo Scientific Chemicals

Magnesium salicylate tetrahydrate, tech., Thermo Scientific Chemicals

Catalog number: 039379.22
100 g, Each
Thermo Scientific Chemicals

Magnesium salicylate tetrahydrate, tech., Thermo Scientific Chemicals

Catalog number: 039379.22
100 g, Each
Quantity
Catalog number: 039379.22
also known as 039379-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
18917-95-8
IUPAC Name
magnesium(2+) bis(2-hydroxybenzoate) tetrahydrate
Molecular Formula
C14H18MgO10
InChI Key
NBQBEWAYWAMLJJ-UHFFFAOYSA-L
SMILES
O.O.O.O.[Mg++].OC1=CC=CC=C1C([O-])=O.OC1=CC=CC=C1C([O-])=O
Appearance (Color)
White to off-white
Form
Powder or crystals or crystalline powder
Assay from Supplier's CofA
≥90.0%

Description

It is used to treat mild to moderate muscular pain. It is also used to treat headaches, general back pain, and certain joint pains like arthritis. It is found in a variety of over-the-counter (OTC) medications as an anti-inflammatory, primarily for back-pain relief.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text