Thermo Scientific Chemicals

Ammonium iron(III) sulfate dodecahydrate, ACS, 98.5-102.0%, Thermo Scientific Chemicals

Catalog number: 039391.36
500 g, Each
Thermo Scientific Chemicals

Ammonium iron(III) sulfate dodecahydrate, ACS, 98.5-102.0%, Thermo Scientific Chemicals

Catalog number: 039391.36
500 g, Each
Quantity
Catalog number: 039391.36
also known as 039391-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7783-83-7
IUPAC Name
iron(3+) ammonium dodecahydrate disulfate
Molecular Formula
FeH28NO20S2
InChI Key
LCPUDZUWZDSKMX-UHFFFAOYSA-K
SMILES
[NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[Fe+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
Assay (unspecified)
98.5-102.0% H₄FeNO₈S₂.1₂H₂O
Impurity content
Ferrous iron (Fe⁺|l) P.T. (limit about 0.001%)
Impurity content
K 0.005%
Impurity content
Na 0.02%
Impurity content
NO₃ 0.01%

Description

Ammonium iron(III) sulfate dodecahydrate is used in dyeing and printing textiles, as analytical lab reagent. It is often employed as an analytical standard, and has been used in a variety of other applications from nanomaterials to general redox reactions. It plays an important role for the inactivation of mycobacteriophage D29 bacteria which is vivale for viable mycobacterium smegmatis and m.tuberculosis. The crystalline ammonium ferrous sulfate has shown useful function as the high absorbed dose and low-temperature dosimeter.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text