(Cyclooctatetraene)iron tricarbonyl, 98%, Thermo Scientific Chemicals
(Cyclooctatetraene)iron tricarbonyl, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(Cyclooctatetraene)iron tricarbonyl, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
0.5 g
2 g
Catalog number 039402.02
also known as 039402-02
Price (USD)/ Each
186.00
-
Add to cart
Quantity:
0.5 g
Request bulk or custom format
Price (USD)/ Each
186.00
Add to cart
(Cyclooctatetraene)iron tricarbonyl, 98%, Thermo Scientific Chemicals
Catalog number039402.02
Price (USD)/ Each
186.00
-
Add to cart
Specifications
Chemical Name or Material(Cyclooctatetraene)iron tricarbonyl
CAS12093-05-9
Health Hazard 1H302+H312+H332
Health Hazard 2GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Health Hazard 3P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
View more
(Cyclooctatetraene)iron tricarbonyl is used as primary and secondary intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only