Thermo Scientific Chemicals

Decamethylferrocene, 99%, Thermo Scientific Chemicals

Catalog number: 039406.06
5 g, Each
Thermo Scientific Chemicals

Decamethylferrocene, 99%, Thermo Scientific Chemicals

Catalog number: 039406.06
5 g, Each
Quantity
Catalog number: 039406.06
Price (USD)
Have Questions?

Chemical Identifiers

CAS
12126-50-0
Molecular Formula
C20H30Fe
InChI Key
VKRKSGODPDKGAD-UHFFFAOYSA-N
SMILES
Cc1c(C)c(C)[c-](C)c1C.C[C-]12[C-]3(C)[C-]4(C)[C-]5(C)[C-]1(C)[Fe]2345
Molecular Weight (g/mol)
326.31
Assay (unspecified)
> 98.5%
Appearance (Color)
Yellow - orange crystals

Description

Decamethylferrocene is used as a superior internal reference redox standard to ferrocene in voltammetric and amperometric titrations. It is used as photocathode for the detection of barium fluoride scintillation light.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text