Thermo Scientific Chemicals

Bis(1,5-cyclooctadiene)iridium(I) tetrafluoroborate, Thermo Scientific Chemicals

Catalog number: 039414.03
1 g, Each
Thermo Scientific Chemicals

Bis(1,5-cyclooctadiene)iridium(I) tetrafluoroborate, Thermo Scientific Chemicals

Catalog number: 039414.03
1 g, Each
Quantity
Catalog number: 039414.03
also known as 039414-03
Price (USD)
Have Questions?

Chemical Identifiers

CAS
35138-23-9
IUPAC Name
iridium(1+) bis((1Z,5Z)-cycloocta-1,5-diene) tetrafluoroboranuide
Molecular Formula
C16H24BF4Ir
InChI Key
NKIQXDAGMKBVPE-QMDOQEJBSA-N
SMILES
[Ir+].F[B-](F)(F)F.C1C\C=C/CC\C=C/1.C1C\C=C/CC\C=C/1
Assay (unspecified)
95% min

Description

Bis(1,5-cyclooctadiene)iridium(I) tetrafluoroborate is used as a halogenated compound for proteomics research. It is also used as intermediates in pharmaceuticals and chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text