Thermo Scientific Chemicals

Calcium stearate, Thermo Scientific Chemicals

Catalog number: 039423.A7
5 kg, Each
Thermo Scientific Chemicals

Calcium stearate, Thermo Scientific Chemicals

Catalog number: 039423.A7
5 kg, Each
Quantity
Catalog number: 039423.A7
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1592-23-0
IUPAC Name
calcium dioctadecanoate
Molecular Formula
C36H70CaO4
InChI Key
CJZGTCYPCWQAJB-UHFFFAOYSA-L
SMILES
[Ca++].CCCCCCCCCCCCCCCCCC([O-])=O.CCCCCCCCCCCCCCCCCC([O-])=O
Comment
Free fatty acid: 0.5 % max
Comment
Softening point: 145 to 165?C
Ash
9.8 to 10.8%
Loss on Drying
3.5% max

Description

Used as a stabilizer and a lubricant in rigid PVC processing, PVC compounding and moulding, in leather cloth manufacturing. It is used as a lubricant, in iron wire drawing and used as an anti caking agent in detergent Cakes and cosmetics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text