Thermo Scientific Chemicals

Terbium(III) bromide, ultra dry, 99.99% (REO), Thermo Scientific Chemicals

Catalog number: 039425.03
1 g, Each
Thermo Scientific Chemicals

Terbium(III) bromide, ultra dry, 99.99% (REO), Thermo Scientific Chemicals

Catalog number: 039425.03
1 g, Each
Quantity
Catalog number: 039425.03
also known as 039425-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
14456-47-4
IUPAC Name
terbium(3+) tribromide
Molecular Formula
Br3Tb
InChI Key
AZNZWHYYEIQIOC-UHFFFAOYSA-K
SMILES
[Br-].[Br-].[Br-].[Tb+3]
Form
Powder
Residual water
< 100 ppm
Appearance (Color)
White
Total Rare Earth Oxide Impurities
0.01% max.

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Ampouled under argon
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text