Potassium 2-ethylhexanoate hydrate, 95%, Thermo Scientific Chemicals
Potassium 2-ethylhexanoate hydrate, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Potassium 2-ethylhexanoate hydrate, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number 039440.14
also known as 039440-14
Price (USD)/ Each
197.65
Online exclusive
219.00 
Save 21.35 (10%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
197.65
Online exclusive
219.00 
Save 21.35 (10%)
Add to cart
Potassium 2-ethylhexanoate hydrate, 95%, Thermo Scientific Chemicals
Catalog number039440.14
Price (USD)/ Each
197.65
Online exclusive
219.00 
Save 21.35 (10%)
-
Add to cart
Chemical Identifiers
CAS3164-85-0
IUPAC Namepotassium 2-ethylhexanoate
Molecular FormulaC8H15KO2
InChI KeyZUFQCVZBBNZMKD-UHFFFAOYNA-M
SMILES[K+].CCCCC(CC)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Purity≥94.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only